Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Brand: Lenz Laborglasinstrumente™ 03001949
This item is not returnable.
View return policy
Specifications
According to DIN EN ISO 4797 and in addition to standard | |
Borosilicate Glass, Duran Glass | |
Round |
250 mL | |
19/26 NS Socket | |
85 mm |
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint > Capacity/Socket Joint: 250mL-19/26 NS Socket
Spot an opportunity for improvement?Share a Content Correction