Learn More
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03214158
 View more versions of this product
Product Code. 10382751
  • £37.40 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Acc. to DIN 12394
  • With side neck at 20° angle
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • Ground joint features high taper accuracy and excellent tightness
Specifications
Specifications
Acc. to DIN 12394 and in addition to DIN standard
Borosilicate Glass, Duran Glass
Round
500 mL
14/23 NS Side Neck, 29/32 NS Center Neck