Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
Brand:  Hirschmann™ 3310160
 View more versions of this product
Product Code. 13158009
  • £378.00 / Pack of 2
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Schellbach
  • Bang pattern
  • Main point ring graduation
  • With blue graduation
  • Acc. to DIN EN ISO 385
  • Support base
  • Conformity certified, with dated batch identification
Specifications
Specifications
10 mL
0.02 mL
Glass
Lateral
±0.02 mL
Class AS
Borosilicate Glass
Bang, Main Point Ring Graduation, Schellbach Stripes, Socket
Blue
Glass
Microburet
DIN EN ISO 385