Learn More
DWK Life Sciences DURAN™ Drying Tube, bent, with standard ground cone
Filled with a desiccant, the drying tube protects a reaction mixture in a flask from moisture.
£24.35 - £39.70
Product Code | Brand | Joint Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | ||||||
10614182
|
DWK Life Sciences
242620609 |
NS 14/23 |
£24.35
Each |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
10090001
|
DWK Life Sciences
242620909 |
NS 29/32 |
£39.70
Each |
In Stock
Log in to see stock. |
||||||
10400503
|
DWK Life Sciences
242620806 |
NS 24/29 |
£28.50
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
10203221
|
DWK Life Sciences
242620703 |
NS 19/26 |
£26.35
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||||
Description
The drying tube is usually an angled glass tube with a standard ground joint at one end. The other end is open. For filling the drying tube is first stuffed with a little cotton wool, then the drying agent is filled in and finally another piece of cotton wool prevents the drying agent from falling out.
- Tight closure due to standard ground joint
- Bent
- With standard ground socket
- Fired-on, highly durable white print
Specifications
1 Ea. |