Learn More
DWK Life Sciences DURAN™ Erlenmeyer Flask, narrow neck
Due to conical shape, the flasks is used for mixing liquids.
£57.39 - £260.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Media preparation and cultivation; Storing and transport; Mixing several components; Heating; Diluting solutions; Measuring liquid amounts; Size 25 mL without Retrace Code und Sizes 150, 200 and 300 mL non-DIN ISO size |
Autoclavable | Yes |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11702172
|
DWK Life Sciences
212161407 |
25 mL |
£59.72
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11712172
|
DWK Life Sciences
212161707 |
50 mL |
£59.80
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11702182
|
DWK Life Sciences
212162403 |
100 mL |
£152.00
Pack of 10 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
11712182
|
DWK Life Sciences
212163202 |
200 mL |
£68.35
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11722182
|
DWK Life Sciences
212163605 |
250 mL |
£144.00
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11732182
|
DWK Life Sciences
212163905 |
300 mL |
£69.80
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
11742182
|
DWK Life Sciences
212164404 |
500 mL |
£80.55
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11752182
|
DWK Life Sciences
212165409 |
1000 mL |
£184.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11762182
|
DWK Life Sciences
212166302 |
2000 mL |
£260.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
11712412
|
DWK Life Sciences
212166808 |
3000 mL |
£125.00
Pack of 2 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
10189910
|
DWK Life Sciences
212167307 |
5000 mL |
£71.08
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
11669868
|
DWK Life Sciences
212162806 |
125 mL |
£57.39
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
13479118
|
DWK Life Sciences
219902702 |
150 mL |
£77.50
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
12626167
|
DWK Life Sciences
212165306 |
800 mL |
£101.00
Pack of 10 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
Description
Thanks to their exceptional properties, such as excellent chemical resistance, nearly inert behaviour and high usage temperature, the original DURAN™ Erlenmeyer flasks are ideal for use in laboratories. Moreover, the uniform wall thickness, minimal thermal expansion and the resultant high thermal shock resistance are outstanding characteristics. Therefore, DURAN™ Erlenmeyer flasks are particularly suited to use with very high temperatures and fast temperature changes.
- Easy mixing due to conical form
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
Borosilicate Glass (3.3) | |
Media preparation and cultivation; Storing and transport; Mixing several components; Heating; Diluting solutions; Measuring liquid amounts; Size 25 mL without Retrace Code und Sizes 150, 200 and 300 mL non-DIN ISO size | |
Yes |