Learn More
DWK Life Sciences DURAN™ Erlenmeyer Flask, with standard ground joint
Erlenmeyer flasks with standard ground joint allow, among other things, the storage of a solution through the tight closure of the vessel.
£12.90 - £350.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Mixing several components; Diluting solutions; Measuring liquid amounts; Heating; Volume sizes 50 mL (NS 24/29), 100 mL (NS 24/29), 200 mL (NS 29/32), 250 mL (NS 45/40), 300 mL (NS 29/32), 500 mL (NS 45/40) and 1000 mL (NS 45/40) are not according to DIN ISO |
No. per Pack | 10/Pk. |
Autoclavable | Yes |
Product Code | Brand | Capacity (Metric) | Thread Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Thread Size | Price | Quantity & Availability | ||||
11756268
|
DWK Life Sciences
241931906 |
50 mL | NS 29/32 |
£178.54
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15231889
|
DWK Life Sciences
241932405 |
100 mL | NS 19/26 |
£178.00
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15251889
|
DWK Life Sciences
241932705 |
100 mL | NS 29/32 |
£222.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
15261889
|
DWK Life Sciences
241933204 |
200 mL | NS 29/32 |
£214.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15271889
|
DWK Life Sciences
241933607 |
250 mL | NS 24/29 |
£214.00
Pack of 10 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
15281889
|
DWK Life Sciences
241933701 |
250 mL | NS 29/32 |
£277.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
15291889
|
DWK Life Sciences
241933804 |
250 mL | NS 45/40 |
£258.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
11850620
|
DWK Life Sciences
241933907 |
300 mL | NS 29/32 |
£220.00
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15201899
|
DWK Life Sciences
241934603 |
500 mL | NS 29/32 |
£265.00
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15231899
|
DWK Life Sciences
241935608 |
1000 mL | NS 29/32 |
£281.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15241889
|
DWK Life Sciences
241932602 |
100 mL | NS 24/29 |
£201.00
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15211899
|
DWK Life Sciences
241934706 |
500 mL | NS 45/40 |
£277.00
Pack of 10 |
Estimated Shipment: 23-10-2024
Log in to see stock. |
||||
15201889
|
DWK Life Sciences
241931306 |
25 mL | NS 14/23 |
£162.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15221899
|
DWK Life Sciences
241935402 |
1000 mL | NS 24/29 |
£281.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15221889
|
DWK Life Sciences
241932002 |
50 mL | NS 14/23 |
£164.00
Pack of 10 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
10273451
|
DWK Life Sciences
241934406 |
500 mL | NS 24/29 |
£241.49
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15211889
|
DWK Life Sciences
241931803 |
50 mL | NS 24/29 |
£208.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
15241899
|
DWK Life Sciences
241935702 |
1000 mL | NS 45/40 |
£350.00
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10385051
|
DWK Life Sciences
241931709 |
50 mL | NS 19/26 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Due to their large footprint, Erlenmeyer flasks are suitable, for example, for preparing solutions and storing them. The additional grinding allows a tight closure of the vessel.
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white print
- Easy mixing due to conical form
Specifications
Borosilicate Glass (3.3) | |
Mixing several components; Diluting solutions; Measuring liquid amounts; Heating; Volume sizes 50 mL (NS 24/29), 100 mL (NS 24/29), 200 mL (NS 29/32), 250 mL (NS 45/40), 300 mL (NS 29/32), 500 mL (NS 45/40) and 1000 mL (NS 45/40) are not according to DIN ISO | |
10/Pk. | |
Yes |