Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks With Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03021937
Code : A0
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12034098
  • £81.99 / Pack of 10
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
DIN EN ISO 4797
Borosilicate Glass, Duran Glass
NS19/26
100 mL
With scale (approx. volume/mL), with inscription label
64 mm