Learn More
DWK Life Sciences DURAN™ Evaporating Dish, with spout
The wide, open shape of the evaporation dish creates a large liquid surface which favours the evaporation of excess solvents.
Brand:  DWK Life Sciences 213015906
514.40 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11707018
  • £621.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Spout for clean pouring
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Borosilicate Glass (3.3)
10/Pk.
1500 mL
Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field
190 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Evaporating Dish, with spout > 1500 mL