Learn More
DWK Life Sciences DURAN™ Evaporating Dish, with spout

The wide, open shape of the evaporation dish creates a large liquid surface which favours the evaporation of excess solvents.
Brand: DWK Life Sciences 213014404
111.74 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
- Spout for clean pouring
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic

Specifications
Autoclavable | |
Borosilicate Glass (3.3) | |
10/Pk. |
170 mL | |
Evaporation of liquids; Preparation of concentrated solutions; Prevention of solvent condensation on glass walls; Sizes 25, 30, 35 and 45 mL without labelling field | |
95 mm |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
6Hours
15Minutes
49Seconds
8 -
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
81Hours
15Minutes
49Seconds
8New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.