Learn More
DWK Life Sciences DURAN™ Filter Disc Sintered Glass, Porosity 1, edge not fused
A prerequisite for successful work with glass filters is the selection of the right porosity. It is important to select a filter whose largest pore size is slightly below the grain size of the smallest particles still to be separated.
£12.50 - £71.10
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Price | Quantity & Availability | |||||
12759746
|
DWK Life Sciences
251510106 |
10 mm |
£41.75
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
|||||
15262019
|
DWK Life Sciences
251520107 |
20 mm |
£43.40
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
15212039
|
DWK Life Sciences
251540109 |
40 mm |
£63.70
Pack of 10 |
Estimated Shipment: 06-11-2024
Log in to see stock. |
|||||
10329510
|
DWK Life Sciences
251590105 |
90 mm |
£29.35
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
11610009
|
DWK Life Sciences
251565109 |
65 mm |
£20.40
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
11679988
|
DWK Life Sciences
251545107 |
45 mm |
£71.10
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
11669978
|
DWK Life Sciences
251515104 |
15 mm |
£43.05
Pack of 10 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
11659998
|
DWK Life Sciences
251555108 |
55 mm |
£12.50
Each |
Estimated Shipment: 20-09-2024
Log in to see stock. |
|||||
15834508
|
DWK Life Sciences
251535106 |
35 mm |
£53.70
Pack of 10 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
11600019
|
DWK Life Sciences
251600103 |
100 mm |
£62.65
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10010021
|
DWK Life Sciences
251620105 |
120 mm |
£69.95
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
15262029
|
DWK Life Sciences
251530108 |
30 mm |
£49.00
Pack of 10 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10131601
|
DWK Life Sciences
251560102 |
60 mm |
£13.60
Each |
In Stock
Log in to see stock. |
|||||
10111651
|
DWK Life Sciences
251570103 |
70 mm |
£23.70
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10263361
|
DWK Life Sciences
251580104 |
80 mm |
£25.95
Each |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
15252039
|
DWK Life Sciences
251550101 |
50 mm |
£42.85
Pack of 5 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||||
15212029
|
DWK Life Sciences
251525105 |
25 mm |
£44.40
Pack of 10 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
Description
Glass powder with a certain particle size distribution results in a porous, solid body. These bodies are used as filter material in the form of plates and candles. Made of DURAN™ (borosilicate glass 3.3). They are ideally suited for separations with strong acids or alkalis, for example, and thus offer advantages over other materials such as plastic or paper. Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99. Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar.
- Edge non-melted and centered, surfaces untreated
- Successful work through the correct choice of porosity (The largest pore size should be slightly below the grain size of the smallest particles still to be separated).
- Available in 6 different porosity classes according to ISO 4796 as well as ASTM EI28-99.
- Glass articles for further processing
Specifications
Borosilicate Glass (3.3) | |
Suitable for vacuum filtration with a maximum resulting pressure difference of 1 bar |