Learn More
DWK Life Sciences DURAN™ Funnel, with short stem
A funnel enables a clean transfer and filtration of liquids.
Brand:  DWK Life Sciences 213516104
Code : A0
Additional Details : Weight : 0.51000kg
 View more versions of this product
Product Code. 10455651
  • £81.85 / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket
Description
Description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
  • With short stem
  • Due to its shape, it is suitable for liquid-solid filtration using a filter paper
  • Fired-on, highly durable white print
Specifications
Specifications
200 mm
Yes
1 Ea.
Borosilicate Glass (3.3)
Decanting and filtration of liquids; The spillage of substances is avoided; Filtering
Non-DIN/ISO size