missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Gas-washing Bottle, lower part, Drechsel type with NS 29/32, height 200 mm

For cleaning gases.

Brand:  DWK Life Sciences 247143606

Code : NEW

Additional Details : Weight : 3.10000kg

 View more versions of this product

Product Code. 15212009

  • £350.00 / Pack of 10
Estimated Shipment: 03-09-2024
to see stock.
Add to basket

Description

Description

DURAN™ gas washing bottle bases can be completed with various attachments to form complete washing bottles.

  • After turning with NS 29/32, height 200 mm
  • According to DIN 12463
  • Can be completed to complete wash bottles with various attachments
Specifications

Specifications

250 mL
Borosilicate Glass (3.3)
Purification of gases
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers