missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GL 45 Bottle
Click to view available options
DURAN™ GL 45 Bottle; Includes: GL 45 Side Neck, Two GL 45 inlets
Description
- Ideal for the storage and delivery of high-performance liquid chromatography (HPLC) mobile phases and solvents.
- Increasing the number of GL 45 inlets on the bottles adds convenience: One additional side neck allows additional solvent to be added without disturbing the main delivery cap.
- Convenient – Side neck allow solvent filling without main delivery cap removal
- Safer – Capped bottles reduce the risk of volatile organic compounds (VOC) escape and spillage
- Compatible - GL 45 threads fit standard HPLC connection caps
- Manufactured from the material of choice for HPLC solvent bottles: borosilicate 3.3 glass offering a very low inorganic ion extractables profile
- Clear DURAN™ is a borosilicate glass type 3.3 as specified by the international standard ISO 3585, and is classified as a ''Type I, Class A low-expansion borosilicate glass'' as defined by ASTM-E438-1992
- Useful 2 Liter Capacity
- Retrace Code, and certificate for manufacturing lot traceability
- Durable white enamel volume graduations
Spécification
Spécification
Capacity (Metric) | 2000 mL |
Cleaning Procedure | Lab washer compatible |
Color | Clear |
Diameter (Metric) Inner Neck | 30 mm |
For Use With (Application) | Mobile Phase Reservoirs for HPLC, Reagent Supply to Liquid Handling Systems and Analyzers |
Includes | GL 45 Side Neck, Two GL 45 inlets |
Material | Borosilicate Glass (3.3) |
Max. Temperature (Metric) | 500°C |
Quantity | 1 |
Screw Color | Blue |
Afficher plus |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'