missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Screw Connection Cap, GL 45, with 2 or 3 GL 14 ports, PP
Shop All DWK Life Sciences Products
Click to view available options
Description:
Screw connection cap GL 45, PP, 2 ports GL 14
Screw connection cap GL 45, PP, 3 ports GL 14
Description
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
- Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts.
- Available with 2, or 3 threaded outlets (ports).
- Different tubing diameters can be used by changing the GL 14 inserts.
- By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap.
- Fully compatible with all the GL 45 threads and therefore highly versatile.
- 1129751 is also fully compatible with all the GLS 80 threads and therefore highly versatile.
Suitable Accessories:
- 1129814
- 1129815
- 1129816
- 1129817
- 1129818
- 1156292
- 1137799
- 1129819

Specifications
Specifications
Description | Screw connection cap GL 45, PP, 2 ports GL 14 |
For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply |
Material | Polypropylene, PTFE, Silicone |
Quantity | 2/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Screw Connection Cap, GL 45, with 2 or 3 GL 14 ports, PP >
Spot an opportunity for improvement?Share a Content Correction