Learn More
DWK Life Sciences DURAN™ Insert for Screw Cap GL 14, Silicone/PTFE (Screw Connection Cap GL 45 Accessory)
Accessory for 2 or 3 port GL 45 connection cap system
£13.75 - £38.15
Specifications
For Use With (Application) | Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply |
---|---|
Material | Polypropylene, PTFE |
Quantity | 1 Ea. |
Thread Size (Metric) | GL 14 |
Autoclavable | Autoclavable |
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
10399652
|
DWK Life Sciences
1129818 |
ID 6.0 mm |
£38.15
Each |
In Stock
Log in to see stock. ![]() |
||||||
10572763
|
DWK Life Sciences
1129815 |
ID 1.6 mm |
£13.75
Each |
In Stock
Log in to see stock. |
||||||
10412153
|
DWK Life Sciences
1129816 |
ID 3.0 mm |
£36.55
Each |
In Stock
Log in to see stock. |
||||||
10523343
|
DWK Life Sciences
1129817 |
ID 3.2 mm |
£35.75
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
Flexible modular GL 45 connection cap system that accepts four different rigid or semi-rigid tubing diameters (1.6 mm; 3.0 mm; 3.2 mm and 6.0 mm). The caps are available with 2 or 3 GL 14 threaded ports. The different sized inserts form a tight seal around the pass-through tubing. Sterile pressure equalisation is possible through use of 0.2 micron syringe filter. Unused ports can be sealed off with a red GL 14 screw cap.
- Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts
- Available with 2, or 3 threaded outlets (ports)
- Five different tubing diameters (3.2 mm, 6.0 mm, 8.0 mm, 10.0 mm and 12.0 mm) can be used by changing the inserts
- By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap
- Fully compatible with all the GL 45 threads and therefore highly versatile
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Suitable Accessory: 1129814
Specifications
Sterile fluid transfer in and out of lab bottles; Gas transfer in and out of lab bottles; Fluid transfer between lab bottles; Supply of reagents to instruments; Bioreactor supply | |
1 Ea. | |
Autoclavable |
Polypropylene, PTFE | |
GL 14 | |
87 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.