Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers
Brand:  Lenz Laborglasinstrumente™ 05240404
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 10017810
  • £125.00 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Specifications
Cooling Surface: 740cm2
For condensing vapors
400 mm
Duran Glass
29/32
Dimroth Condenser
1 Each
DIN 12591 and in addition to DIN standard