Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for secure and easy leak-tight connection of apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
£7.22 - £15.00
Product Code | Brand | Cone Size | Standard Taper Joint No | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Standard Taper Joint No | Price | Quantity & Availability | ||||
10088450
|
Lenz Laborglasinstrumente™
01311414 |
NS 14/23 | GL 14 |
£7.22
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||||
10597741
|
Lenz Laborglasinstrumente™
01311419 |
NS 19/26 | GL 14 |
£9.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
10692411
|
Lenz Laborglasinstrumente™
01311819 |
NS 19/26 | GL 18 |
£10.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
10324001
|
Lenz Laborglasinstrumente™
01312519 |
NS 19/26 | GL 25 |
£11.00
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||||
10363901
|
Lenz Laborglasinstrumente™
01311429 |
NS 29/31 | GL 14 |
£11.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
10708931
|
Lenz Laborglasinstrumente™
01311424 |
NS 24/29 | GL 14 |
£11.00
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
10160121
|
Lenz Laborglasinstrumente™
01312529 |
NS 29/32 | GL 25 |
£12.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
10405801
|
Lenz Laborglasinstrumente™
01311824 |
NS 24/29 | GL 18 |
£11.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
10632421
|
Lenz Laborglasinstrumente™
01311829 |
NS 29/32 | GL 18 |
£9.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10002944
|
Lenz Laborglasinstrumente™
01313229 |
NS 29/32 | GL 32 |
£15.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
||||
Description
- Acc. to DIN 12257
- Available in a range of socket and thread sizes
- Minimized need for grease
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
1 Ea. | |
Duran Glass |