Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Provides for quick and easy leak-tight connections to apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 01311814
This item is not returnable.
View return policy
Description
- Acc. to DIN 12257
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifications
DIN 12257 | |
Borosilicate Glass (3.3) | |
1 Pack | |
Screwthread Tubes |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone > 18GL Thread; 14/23 NS cone
Spot an opportunity for improvement?Share a Content Correction