Learn More
DWK Life Sciences DURAN™ GLS 80™ High Temperature Pouring Ring
Click to view available options
White
Description
The material used for this GLS 80 closure is a special compound based on polyarylsulphone. As a result, the chemical, thermal and mechanical properties of the material are improved compared to polypropylene. Thanks to the shortened triple thread, the DURAN™ GLS 80™ bottle can be opened or closed with only a three-quarter turn. The seal, which is coated on both sides with PTFE, ensures the bottle can be tightly sealed (cap liner: Platinum-cured silicone). A matching PTFE pouring ring is also available, permitting clean, drip-free use.
- PSU caps and PTFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components
- Screw cap seal with PTFE faced cap liner for efficient sealing
- The matching PTFE pouring ring permits drip-free pouring and clean, safe working
Specifications
Description | Pouring ring |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping |
Material | PSU, PBT, ETFE |
Quantity | 5/Pk. |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |