Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber, GLS 80™ Thread, USP /EP (3.2.1) Light Transmission, Graduated
Wide mouth, multifunctional glass laboratory bottles with UV protection.
Brand: DWK Life Sciences 218663604
Code : A0
Additional Details : Weight : 3.20000kg
Description
DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
Specifications
250 mL | |
Without screw cap and pouring ring from PP (blue) | |
10/Pk. | |
Autoclavable | |
95 mm |
Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir | |
Borosilicate Glass (3.3) | |
GLS 80 | |
Polypropylene |