missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle Only, Wide Mouth, Amber, GLS 80™ Thread, USP /EP (3.2.1) Light Transmission, Graduated

Wide mouth, multifunctional glass laboratory bottles with UV protection.

Brand:  DWK Life Sciences 1178429

388.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 4.50000kg

 View more versions of this product

Product Code. 15971371

  • £485.00 / Pack of 10
Estimated Shipment: 04-09-2024
to see stock.
Add to basket

Description

Description

DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.

  • UV protection up to approx. 500 nm due to external colouration. The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
  • Easy access to the bottle contents due to the wide diameter of the bottle neck
  • Special thread design for rapid opening and closing in less than a turn
  • Optimum filling and pouring and particularly easy cleaning
  • Spoons, spatulas and tweezers can be manipulated with ease
  • Even large-volume funnels can be inserted into the bottles without any difficulty
  • Stability due to the large base area
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
Specifications

Specifications

500 mL
Without screw cap and pouring ring from PP (blue)
10/Pk.
Autoclavable
101 mm
Autoclaving of of light sensitive media and reagents; Preparation of light sensitive media and reagents; Sample collection of light sensitive materials; Culturing vessel; Storage of light sensitive media and reagents; Transport and shipping of light sensitive media and reagents; Liquid handling reservoir
Borosilicate Glass (3.3)
GLS 80
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers