Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, GLS 80™ Thread, Graduated, Bottle Only
Wide mouth, multifunctional glass laboratory bottles.
£8.00 - £684.00
Specifications
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
---|---|
Includes | Without screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15951371
|
DWK Life Sciences
1178424 |
1000 mL |
£212.00
Pack of 10 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
10061263
|
DWK Life Sciences
1178425 |
2000 mL | N/A | ||||||
10543135
|
DWK Life Sciences
1178392 |
500 mL | N/A | ||||||
11689928
|
DWK Life Sciences
218606904 |
3500 mL |
£123.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
10113343
|
DWK Life Sciences
1178426 |
5000 mL |
£170.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
10376453
|
DWK Life Sciences
1178427 |
10,000 mL |
£318.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
10659353
|
DWK Life Sciences
1178428 |
20,000 mL |
£684.00
Each |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
12646167
|
DWK Life Sciences
218603607 |
250 mL |
£88.05
Pack of 10 |
Estimated Shipment: 23-12-2024
Log in to see stock. |
|||||
Description
DURAN™ wide mouth GLS 80™ laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ wide mouth GLS 80™ are supplied complete with blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
Specifications
Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir | |
Borosilicate Glass (3.3) | |
Autoclavable |
Without screw cap and pouring ring from PP (blue) | |
GLS 80 | |
Polypropylene |