Learn More
DWK Life Sciences DURAN™ GLS 80™ Laboratory Bottle, Wide Mouth, Clear, Protect coated, GLS 80™ Thread, Plastic safety coated, Graduated
Wide mouth, multifunctional glass laboratory bottles with protective safety coating.
£248.00 - £850.00
Specifications
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
---|---|
Includes | With screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Closure Material | Polypropylene |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11796988
|
DWK Life Sciences
1160152 |
500 mL | N/A | ||||||
11706998
|
DWK Life Sciences
1160163 |
1000 mL | N/A | ||||||
11716998
|
DWK Life Sciences
1160164 |
2000 mL | N/A | ||||||
10295203
|
DWK Life Sciences
1160165 |
5000 mL | N/A | ||||||
12666167
|
DWK Life Sciences
218653652 |
250 mL | N/A | ||||||
11619938
|
DWK Life Sciences
218656958 |
3500 mL | N/A | ||||||
Description
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.
- The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 ml and less)
- UV protection up to approx. 380 nm due to external coating
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Special thread design for rapid opening and closing in less than a turn
- Optimum filling and pouring and particularly easy cleaning
- Spoons, spatulas and tweezers can be manipulated with ease
- Even large-volume funnels can be inserted into the bottles without any difficulty
- Stability due to the large base area
- Easy-to-read scale with graduation marks to estimate the content
Specifications
Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir | |
Borosilicate Glass (3.3) | |
Polypropylene |
With screw cap and pouring ring from PP (blue) | |
Autoclavable |