Learn More
DWK Life Sciences DURAN™ GLS 80™ O-Ring Sealing Gasket, Black EPDM rubber, GLS 80™ Wide Mouth
Optional sealing gasket for GLS 80™ bottles.
Brand:  DWK Life Sciences 1152913
Code : NEW
Additional Details : Weight : 0.01800kg
Product Code. 12826573
  • £9.80 / Pack of 5
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
EPDM O-Ring gasket improves sealing between GLS 80™ stirred reactor and the glass bottles.
  • Improved sealing with GLS 80™ bottles
Specifications
Specifications
Liquid tight seal
5/Pk.
EPDM
78 mm