Learn More
DWK Life Sciences DURAN™ GLS 80™ Production and Storage Bottle, Carboys, Clear, Protect coated
Description
These larger sized bottles / carboys are ideal for bulk storage and handling of both liquid and solid intermediates and final formulations. Manufactured from Type 1 borosilicate 3.3 glass for durable performance and resistance to thermal stress. The glass conforms to American (USP), European (EP) and Japanese pharmacopoeia (JP) standards making the carboys ideal for pharmaceutical production applications. Manufactured with thickened, uniform side walls for higher mechanical strength. Retrace Code for batch traceability and conformance certification. Manufactured from inorganic materials (Certified BSE / TSE free). Suitable for high temperature sterilization, depyrogenation or autoclaving. Feature large, permanent, easy-to-read, white enamel graduations marks. External polyurethane protect coating for enhanced scratch resistance. Available with customized logos, identification labeling or graduations Supplied without screw cap or pouring ring, but can be used in conjunction caps and connector systems. Typical applications: Flat robust base is ideal for mixing processes with large magnetic stir bars.
- The coating provides scratch protection
- Thick-walled and therefore mechanically very resistant Manufactured from inorganic materials (Certified BSE/TSE free)
- Manufactured from inorganic materials (Certified BSE/TSE free)
- The flat, robust base allows the use of large and heavy stir bars
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Optimum filling and pouring and particularly easy cleaning
Specifications
Autoclavable | Autoclavable |
Capacity (Metric) | 20,000 mL |
Diameter (Metric) Outer | 289 mm |
For Use With (Application) | Pharmaceutical manufacturing; API mixing processes with large magnetic stir bars; Autoclaving of media and reagents; Preparation of media and reagents |
Material | Borosilicate Glass (3.3), Polyurethane |
Neck Style | Thread Size GLS 80 |
No. per Pack | 1 Ea. |