missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GLS 80™ Screw Cap, Blue PP, Linerless Lip Seal
32.51 GBP valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Blue
Description
Permits opening and closing of the DURAN™ GLS 80™ bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
- Triple thread permits user friendly opening and closing with only a three-quarter turn
- Cap offer a linerless, integral lip seal
- Pouring ring for drip-free pouring and clean, safe working
Specifications
Description | Screw cap |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping |
Material | Polypropylene |
Quantity | 10/Pk. |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |
Diameter (Metric) | 87 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'