Learn More
DWK Life Sciences DURAN™ GLS 80™ screw cap with four ports
Product Code. 15710239 Shop All DWK Life Sciences Products
Blue, Gray
Click to view available options
Blue, Gray
Flexible GLS 80™ cap connection system for use with pass through semi-rigid tubing.
Flexible modular liquid or gas transfer GLS 80 cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap
  • The flexible modular system allows an individual combination of the different components according to requirements and application
  • Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts
  • Sterile pressure equalisation is possible by using the syringe filter
  • Unused ports can be sealed off with a blank GL 18 screw cap
  • Fully compatible with all the GLS 80 threads and therefore highly versatile
TRUSTED_SUSTAINABILITY
Specifications
Description DURAN GLS 80 screw cap with four ports (GL 18 thread) Blue and grey Polypropylene with white EPDM O-ring
For Use With (Application) Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments; Photo-bioreactor
Material Polypropylene, PTFE, EPDM
Quantity 2/Pk.
Thread Size (Metric) GLS 80
Autoclavable Autoclavable
Diameter (Metric) 87 mm
Product Content Correction
Product Title
DWK Life Sciences DURAN™ GLS 80™ screw cap with four ports >