missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ GLS 80™ Wide Mouth Bottle Connection System, Screw Cap GLS 80™, Central Standard Taper 29/32, 4-Ports, GL 18 Thread
Click to view available options
2 x GL 14 thread, 2 x GL 18 thread, central ST 29/32 ground joint
Description
Flexible modular liquid or gas transfer GLS 80 cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
- The flexible modular system allows an individual combination of the different components according to requirements and application
- Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts
- Sterile pressure equalisation is possible by using the syringe filter
- Unused ports can be sealed off with a blank GL 18 screw cap
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Specifications
Diameter (Metric) | 87 mm |
Description | Accessories GL 14 |
For Use With (Application) | Fluid transfer; Supply of liquid into bioreactors; Supply of reagents to instruments |
Material | Polypropylene, PTFE |
Quantity | 2/Pk. |
Thread Size (Metric) | GLS 80 |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'