missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
Click to view available options
Thread Size (Metric):
GL 14
GL 18
GL 25
GL 32
GL 45
Description
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
- Excellent chemical resistance due to PTFE coating
- Cap liner forms a tight seal with bottle Peroxide catalysed silicone
- Peroxide catalysed silicone
- Suitable for PBT screw caps and tamper-evident screw caps
- Fully compliant with current international food contact regulations
Specifications
Specifications
Color | Red |
Description | Screw cap |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping |
Material | PBT, ETFE, Fiberglass |
Quantity | 10/Pk. |
Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
Thread Size (Metric) | GL 25 |
Liner Material | PTFE-coated Silicone |
Autoclavable | Autoclavable |
Closure Liner | PTFE/Silicone |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'