missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Laboratory Bottle, Protect coated, Amber, with DIN 168-1 Thread, Plastic safety coated, USP / EP (3.2.1) Light Transmission

Multifunctional glass laboratory bottles with UV protection and a protective safety coating.

Brand:  DWK Life Sciences 1173548

Code : NEW

Additional Details : Weight : 3.50000kg

 View more versions of this product

Product Code. 11699838

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, ambered, glass that is then coated with a PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. The amber ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.

  • Standardized GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 mL and less)
  • UV protection up to approx. 500 nm due to external coloration
  • The only external application of the color ensures that internal surfaces have standard DURAN™ glass properties
  • Easy-to-read scale with graduation marks to estimate the content
  • Sizes 10 and 25 mL with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Specifications

Specifications

10,000 mL
Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel
Borosilicate Glass (3.3)
GL 45
227 mm
Polyurethane
Without screw cap and pouring ring
1 Ea.
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers