missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated

Código de producto. 11341465
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL

Multifunctional glass laboratory bottles with square space saving shape.

DURAN™ Square laboratory bottles have a space saving shape that offers the benefit of close and efficient packing. With up to a 20 % saving of space compared to traditional round bottles. Ideal for the shipping and storage of high-demand materials. For reagent supply to instruments where bench space is at a premium. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ square bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.

  • Ergonomic handling, high stability and good stackability due to angular shape
  • Saves space and costs in autoclaves, drying chambers, ovens, storage or washers
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY

Especificaciones

Capacity (Metric) 1000 mL
For Use With (Application) Liquid handling reservoir for instruments; Transport and shipping; Sample collection; Storage of media and reagents; Archival storage of samples; Preparation of media and reagents; Culturing vessel
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Autoclavable Autoclavable
Closure Material Polypropylene
Diameter (Metric) 94 mm
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'