Learn More
WTW™ DURAN™ Measuring Bottle
WTW offers a wide variety of measuring bottles and measuring vessels with adapters to measure BSB, soil respiration, degradeability as per OECD 301F, fermentation tests, biogas and much more. They have all be tested in their respective applications and therefore support accurate measurements.
Brand: WTW™ 209107
Code : NEW
Additional Details : Weight : 1.00000kg
Specifications
Measuring Bottle Duran | |
2 Nozzles with 2 x VS25 Screwing Sets for Nozzle, 1 x GK 600 Rubber Sleeve, 2 x SEP25 Septum for Nozzles, 2 x STH25 Support Sleeves for Nozzle | |
500mL |