missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks

Product Code. 11915956
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Diameter (Metric) Outer:
31 mm
45 mm
51 mm
64 mm
85 mm
105 mm
131 mm
Standard Taper Joint:
NS 14/23
NS 29/32
Unit Size:
Each
Pack of 10

Product Code. 11915956

Brand: Lenz Laborglasinstrumente™ 03002958

Only null left
Add to basket
Add to basket

Manufactured from Duran™ glass. Lenz Laborglasinstrumente™ Duran™ Medium-neck Round Bottom Flasks include a conical ground joint.

  • Manufactured from Duran™ glass
  • Complies with DIN EN ISO 4797 standards
  • Includes conical ground joint

Specifications

Certifications/Compliance DIN EN ISO 4797
Capacity (Metric) 500 mL
Material Borosilicate Glass, Duran Glass
Neck Style Medium
Standard Taper Joint NS 29/32
Bottom Shape Round
Diameter (Metric) Outer 105 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.