missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT
PBT screw cap with an open top and high temperature and chemical resistance.
£30.80 - £184.00
Specifications
Color | Red |
---|---|
For Use With (Application) | Aseptic reagent delivery and culture sampling using a syringe and needle |
Material | PBT |
Quantity | 10/Pk. |
Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size (Metric) | Price | Quantity & Availability | ||||||
10434301
|
DWK Life Sciences
292271007 |
GL 45 |
£184.00
Pack of 10 |
In Stock
Log in to see stock. ![]() |
||||||
11767198
|
DWK Life Sciences
292270602 |
GL 18 |
£35.55
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11757008
|
DWK Life Sciences
292270508 |
GL14 |
£30.80
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11737118
|
DWK Life Sciences
292270902 |
GL 25 |
£46.70
Pack of 10 |
In Stock
Log in to see stock. |
||||||
11787118
|
DWK Life Sciences
292270808 |
GL 32 |
£65.10
Pack of 10 |
In Stock
Log in to see stock. |
||||||
Description
Open topped cap with central aperture suitable for use with bromobutyl stoppers, silicone piercing septa or glassware connections. More chemically resistant than PP cap.- Compatible with GL bottle threads
- Red PBT offers high temperature and chemical resistance
- Glass reinforced PBT offers high strength
Specifications
Red | |
PBT | |
Polybutylene Terephthalate (PBT) with glass fiber | |
Open Top Screw Cap |
Aseptic reagent delivery and culture sampling using a syringe and needle | |
10/Pk. | |
Autoclavable | |
PBT with Glass Fiber |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Open Topped Screw Cap, Central Aperture, Red PBT