Learn More
DWK Life Sciences DURAN™ Original Laboratory Bottle, Amber, with DIN 168-1 Thread, USP / EP (3.2.1) Light Transmission, Graduated, Bottle Only
The gold standard for multifunctional glass laboratory bottles.
£183.00 - £886.00
Specifications
For Use With (Application) | Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel |
---|---|
Includes | Without screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10272181
|
DWK Life Sciences
218067309 |
5000 mL |
£239.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
13137246
|
DWK Life Sciences
218066304 |
2000 mL |
£840.00
Pack of 10 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11760864
|
DWK Life Sciences
218065402 |
1000 mL |
£550.00
Pack of 10 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11730854
|
DWK Life Sciences
218061409 |
25 mL |
£270.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11720864
|
DWK Life Sciences
218061709 |
50 mL |
£247.00
Pack of 10 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
11730864
|
DWK Life Sciences
218062405 |
100 mL |
£456.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11740864
|
DWK Life Sciences
218063607 |
250 mL |
£325.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
13121764
|
DWK Life Sciences
218064406 |
500 mL |
£886.00
Pack of 10 |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10442961
|
DWK Life Sciences
218068605 |
10,000 mL |
£409.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10232231
|
DWK Life Sciences
218069104 |
20,000 mL |
£707.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10161881
|
DWK Life Sciences
218068802 |
15,000 mL |
£447.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11766288
|
DWK Life Sciences
218062902 |
150 mL |
£284.00
Pack of 10 |
Estimated Shipment: 18-03-2025
Log in to see stock. |
|||||
11689918
|
DWK Life Sciences
218065102 |
750 mL |
£474.00
Pack of 10 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11639928
|
DWK Life Sciences
218066904 |
3500 mL |
£183.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
13427119
|
DWK Life Sciences
218060807 |
10 mL |
£214.00
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
Description
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, amber, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The amber bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original amber bottles are supplied with or without iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- UV protection up to approx. 500 nm due to external colouration
- The only external application of the colour ensures that internal surfaces have standard DURAN™ glass properties
- Very durable and chemically resistant amber colour
- Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
- Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required
Specifications
Preparation of light-sensitive materials; Storage of light-sensitive materials; Transport and shipping of light-sensitive materials; Autoclaving of media and reagents; Culturing vessel | |
Without screw cap and pouring ring from PP (blue) | |
Borosilicate Glass (3.3) | |
Autoclavable |