missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated Product Code.: 12347178
Add to basket

DWK Life Sciences DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated

Product Code. 12347178 Shop All DWK Life Sciences Products
1000 mL
Click to view available options
Capacity (Metric):
10 mL
10,000 mL
100 mL
1000 mL
15,000 mL
150 mL
2 L
20,000 mL
25 mL
25,000 mL
250 mL
3500 mL
50 mL
500 mL
5000 mL
750 mL

Product Code. 12347178

Brand: DWK Life Sciences 218015455

Edge
Add to basket
Add to basket

The gold standard for multifunctional glass laboratory bottles.

DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.

  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Retrace Code, and certificate for manufacturing lot traceability
  • Fired-on, highly durable white print
  • Suitable for long-term storage
  • Easy-to-read scale with graduation marks to estimate the content
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir
Includes With screw cap and pouring ring from PP (blue)
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Autoclavable Autoclavable
Diameter (Metric) 101 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.