Learn More
DWK Life Sciences DURAN™ Original Laboratory Bottle, Clear, with DIN 168-1 Thread, Graduated, Bottle Only
Use promo code "21690" to get your promotional price.
Description
DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Retrace Code, and certificate for manufacturing lot traceability
- Fired-on, highly durable white print
- Suitable for long-term storage
- Easy-to-read scale with graduation marks to estimate the content
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity (Metric) | 2000 mL |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
Includes | Without screw cap and pouring ring from PP (blue) |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Diameter (Metric) | 136 mm |