missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Original Laboratory GL Bottle Pouring Ring
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 32
GL 45
Color:
Blue
Gray
Green
Yellow
Description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
- Cap with integral lip seal provides effective sealing for short term storage
- Pouring ring for drip-free pouring
- The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
- Fully compatible with all the GL 45 threads and therefore highly versatile

Specifications
Specifications
Color | Blue |
Description | Pouring ring |
For Use With (Application) | Autoclaving media and reagents; Short term sample and media storage; color coding and identification |
Material | Polypropylene |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
New Offer Save Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Original Laboratory GL Bottle Pouring Ring >
Spot an opportunity for improvement?Share a Content Correction