missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP
Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 25
GL 32
GL 45
Color:
Blue
Gray
Green
Yellow
Description
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
- Cap with integral lip seal provides effective sealing for short term storage
- Pouring ring for drip-free pouring
- The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
- Fully compatible with all the GL 45 threads and therefore highly versatile

Specifications
Specifications
Color | Blue |
Description | Screw cap |
For Use With (Application) | Autoclaving media and reagents; Short term sample and media storage; color coding and identification |
Material | Polypropylene |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 32 |
Autoclavable | Autoclavable |
Diameter (Metric) | 40 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Original Laboratory GL Bottle Screw Cap, Linerless Lip Seal, PP >
Spot an opportunity for improvement?Share a Content Correction