Learn More
DWK Life Sciences DURAN™ Petri Dish, pressed
Petri dishes are used for the cultivation of bacteria, yeasts, plant and animal cells on solid media.
Brand:  DWK Life Sciences 217544803
87.76 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10796771
  • £130.00 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Cultivation of microorganisms can take place on a gel-like nutrient medium, which is placed in a flat, round, transparent dish with lid (the so-called Petri dish). Due to the solid gel, mixing of the microorganisms is prevented and larger cell culture accumulations (colonies) can grow. DURAN™ Petri dishes are manufactured from borosilicate glass 3.3 using a special manufacturing process. They allow an even distribution of the culture medium and guarantee a distortion-free view. The lids of the DURAN™ Petri dishes rest directly on the lower part and thus prevent turbulence of the protruding air space. This is well suited for reactions with long incubation times, at high temperatures or with sensitive culture media.
  • Facilitated fractionated dilution smear by division (into 2, 3 or 4 parts)
  • Available half-, three- and four-sectional or without sections
  • Pressed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Without Sections
10/Pk.
Borosilicate Glass (3.3)
Cultivation on different culture media at the same time under the same conditions; Growth and cultivation of microorganisms; Cultivation on solid gel culture medium; Microscopy of microorganisms; cell culture
100 x 20 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences DURAN™ Petri Dish, pressed > Without section, diameter 100 x 20 mm