Learn More
DWK Life Sciences DURAN™ Pipeline Filter
A pipeline filter is used to separate solids from a gas phase.
Brand:  DWK Life Sciences 258550404
Code : NEW
Additional Details : Weight : 0.02900kg
 View more versions of this product
Product Code. 10131891
  • £60.70 / Each
Estimated Shipment: 13-09-2024
to see stock.
Add to basket
Description
Description
In addition to solid-liquid filtration, filtration or separation of reaction mixtures from the gas phase is also possible. If, for example, a pipeline filter is installed in a hose system, these filters solid residues from the gas phase and thus purifies it.
  • By installing the filter in a pipe system, the purification of a carrier gas is possible
  • Fired-on, highly durable white print
Specifications
Specifications
Porosity 4
In-line filtration of gas lines to remove solid impurities (e.g. dust); Qualitative inorganic analysis and preparative chemistry; Filtration at reduced pressure; Filtering
30 mm
Borosilicate Glass (3.3)
1/Pk.