missing translation for 'onlineSavingsMsg'
Learn More
DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated Product Code.: 11716888
Estimated Shipment: 24-03-2025
to see stock.
Add to basket

DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated

Product Code. 11716888 Shop All DWK Life Sciences Products
1000 mL
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy

Product Code. 11716888

Brand: DWK Life Sciences 1127078

Estimated Shipment: 24-03-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Glass laboratory bottles with superior thermal and chemical resistance.

The DURAN™ premium GL 45 laboratory bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The premium bottles are ideal for high temperature applications: the thermal shock resistance of the glass bottle is certified for a 160°C differential, and is combined with a high temperature fluoropolymer (PFA) screw cap and pouring ring that are suitable for use up to 200°C. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature a double graduated scale for filling volume and a reversed scale for pouring. The marking field and Retrace Code for manufacturing lot traceability are printed in permanent white enamel.

  • Additional graduation marks and an opposite scaling (accurate to ± 5 %) simplify the reading of the filling level as well as the quantity withdrawn
  • Colourant free product
  • Reliable sealing due to PTFE-coated cap liner
  • TÜV tested thermal shock resistance of 160 K
  • Complete with PFA pouring ring and cap
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage and transport of media and reagents; Shipping
Includes With premium screw cap and pouring ring
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Thread Size (Metric) GL 45
Autoclavable Autoclavable
Closure Material PFA/PFA, PTFE/Silicone Layer
Diameter (Metric) 101 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences DURAN™ Premium Bottle, Clear, with DIN 168-1 Thread, GL 45, Graduated >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.