Learn More
DWK Life Sciences™ DURAN™ protect+ Graduated Laboratory Bottles with DIN 168-1 Thread
Description
DURAN™ GL ISO bottles have become ideal multi-functional laboratory bottles since their introduction in 1972. The protect+ safety-coated version of the bottles combines the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 on the inside and a protective safety plastic layer on the outside. The bottles are molded from high-purity, clear glass or externally ambered clear glass that is then coated with a translucent polyamide PA 12 coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
A combination of high-performance materials and robust design provides a cost-effective usable life. The bottles feature an easy-to-read graduated volume scale. There is a Retrace Code printed on each bottle for manufacturing lot traceability. The ambered DURAN™ glass bottle versions ensure that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards. All the protect+ coated bottles are supplied with push-on protective covers to protect the inside of the bottles from contamination during distribution. Matching screw closures and pouring rings can be ordered separately. Available in a wide range of nominal bottle capacities from 25 mL to 5 L, and neck sizes GL 25 to GLS 80™ wide mouth.
- Manufactured from polyamide-coated DURAN™ borosilicate glass 3.3
- DURAN™ is a neutral glass, classified as a Glass Type I according to USP, EP (3.2.1) and JP, and meets ASTM E438, Type I, Class A requirements
- Reduce the hazards of broken glass and liquid splashes in the event of breakage
- External polymer coating provides scratch, leak* and splinter* protection (*only applies to bottles 2000 mL or less)
- Polyamide 12 safety coating has a high chemical resistance
- Protective coating provides an enhanced safer grip
- Coating is Cleanroom certified Fraunhofer IPA TESTED DEVICE™
- Certified FDA and EU food contact safe coating material
- Safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs) powders
- Supplied with push-on dust cover
- Coating process does not use any volatile organic compounds (VOCs) or isocyanates
Specifications
Specifications
Capacity (Metric) | 100 mL |
Capacity (English) | 3.4 oz. |
Color | Clear |
For Use With (Application) | Preparation of media and reagents, safe storage and shipping of bulk, high-potency active pharmaceutical ingredients (HPAPIs), sample or harvest collection container, culturing vessel, storage of media and reagents, transport and shipping, Liquid handling reservoir |
Material | Borosilicate Glass (3.3) |
Quantity | 10 Bottles |
Sterility | Non-sterile |
Weight (Metric) | 0.159 kg |
Thread Size (Metric) | GL 45 |
Certifications/Compliance | CoC |
Show More |