missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only
DURAN™ Protect Laboratory Bottles are clear, multifunctional borosilicate glass laboratory bottles with a protective safety coating, DIN Threads, and graduations.
£172.00 - £1435.00
Specifications
Coating | Polyurethane |
---|---|
Color | Clear |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir |
Includes | Without screw cap and pouring ring |
Certifications/Compliance | ASTM E438, Type I, Class A |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11762312
|
DWK Life Sciences
218052404 |
100 mL | N/A | |||||||
11782312
|
DWK Life Sciences
218054405 |
500 mL | N/A | |||||||
11792312
|
DWK Life Sciences
218055401 |
1000 mL | N/A | |||||||
10051192
|
DWK Life Sciences
1092676 |
25 mL |
£178.00
Pack of 10 |
Estimated Shipment: 15-04-2025
Log in to see stock. |
||||||
10041192
|
DWK Life Sciences
1092677 |
50 mL | N/A | |||||||
11772312
|
DWK Life Sciences
218053606 |
250 mL |
£178.00
Pack of 10 |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||||
10098060
|
DWK Life Sciences
218057308 |
5000 mL |
£723.00
Each |
In Stock
Log in to see stock. |
||||||
11702322
|
DWK Life Sciences
218056303 |
2000 mL | N/A | |||||||
10625131
|
DWK Life Sciences
218058604 |
10,000 mL | N/A | |||||||
11659828
|
DWK Life Sciences
218059103 |
20 L | N/A | |||||||
11689898
|
DWK Life Sciences
218052901 |
150 mL | N/A | |||||||
11629908
|
DWK Life Sciences
218055101 |
750 mL | N/A | |||||||
11669908
|
DWK Life Sciences
218056903 |
3500 mL | N/A | |||||||
13197216
|
DWK Life Sciences
218058801 |
15,000 mL | N/A | |||||||
13417119
|
DWK Life Sciences
218050806 |
10 mL |
£194.00
Pack of 10 |
Estimated Shipment: 23-04-2025
Log in to see stock. |
||||||
Description
- DURAN™ Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972.
- The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside.
- The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage.
- A combination of high-performance materials and robust ISO 4796-1 design provides a long working life.
- The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel.
- Permanent Retrace Code on each bottle for manufacturing lot traceability.
- The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134°C.
- The coating provides scratch, leak* and splinter* protection (* only applies to bottles 5000 mL and less).
- Highly resistance, long service-life coating.
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out.
- UV protection up to approx. 380 nm due to external coating.
- Easy-to-read scale with graduation marks to estimate the content.
- Fired-on, highly durable white ceramic print.
- Sizes 10 and 25 mL with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Specifications
Polyurethane | |
Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping; Liquid handling reservoir | |
ASTM E438, Type I, Class A | |
Reusable | |
Narrow | |
Safety-coated Bottle |
Clear | |
Without screw cap and pouring ring | |
No | |
Yes | |
Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Protect Clear Borosilicate Glass Laboratory Bottles with Plastic Safety Coating, DIN Thread, Graduations, Bottle Only