Learn More
DWK Life Sciences DURAN™ PURE Bottle, Amber, with DIN Thread
BIOPHARMA
A range of high-quality bottles with light protection and closures, developed for the needs of the pharmaceutical and biotech industry
£299.52 - £24535.00
Specifications
For Use With (Application) Manufacturing of light-sensitive pharmaceutical drugs and biotech products; Storage and transport of light-sensitive APIs, bulk drug intermediates, ingredients and media
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
View More Specs
Products 18
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15565360
BIOPHARMA
View Documents
DWK Life Sciences
818061406
25 mL
£24535.00
Pack of 940
In Stock
to see stock.
Add to basket
15310604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818061406
25 mL
£365.00
Pack of 10
In Stock
to see stock.
Add to basket
15320604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818062402
100 mL
£529.00
Pack of 10
In Stock
to see stock.
Add to basket
15575360
BIOPHARMA
View Documents
DWK Life Sciences
818062402
100 mL
£16055.00
Pack of 540
In Stock
to see stock.
Add to basket
15585360
BIOPHARMA
View Documents
DWK Life Sciences
818063604
250 mL
£11900.00
Pack of 280
In Stock
to see stock.
Add to basket
15330604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818063604
250 mL
£591.00
Pack of 10
In Stock
to see stock.
Add to basket
15336798
BIOPHARMA
View Documents
DWK Life Sciences
818064403
500 mL
£19005.00
Pack of 360
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15340604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818064403
500 mL
£564.08
Pack of 10
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15350604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818065408
1000 mL
£711.36
Pack of 10
In Stock
to see stock.
Add to basket
15346798
BIOPHARMA
View Documents
DWK Life Sciences
818065408
1000 mL
£14020.00
Pack of 210
In Stock
to see stock.
Add to basket
15356798
BIOPHARMA
View Documents
DWK Life Sciences
818066301
2000 mL
£12030.00
Pack of 120
In Stock
to see stock.
Add to basket
15360604
BIOPHARMA
View Documents Promotion Details
DWK Life Sciences
818066301
2000 mL
£1005.00
Pack of 10
In Stock
to see stock.
Add to basket
15896931
View Documents Promotion Details
DWK Life Sciences
818067306
5000 mL
£299.52
Each
In Stock
to see stock.
Add to basket
15508441
BIOPHARMA
View Documents
DWK Life Sciences
818067306
5000 mL
£13325.00
Pack of 48
In Stock
to see stock.
Add to basket
15518441
BIOPHARMA
View Documents
DWK Life Sciences
818068602
10,000 mL
£12330.00
Pack of 30
In Stock
to see stock.
Add to basket
15806941
BIOPHARMA
View Documents
DWK Life Sciences
818068602
10,000 mL
£411.00
Each
In Stock
to see stock.
Add to basket
15528441
BIOPHARMA
View Documents
DWK Life Sciences
818069101
20,000 mL
£8770.00
Pack of 12
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
15816941
BIOPHARMA
View Documents
DWK Life Sciences
818069101
20,000 mL
£733.00
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
Description
Description
High-quality Type I borosilicate glass bottles GL 25 / GL 45, amber coloured from 25 ml – 20 l, developed for the needs of the pharmaceutical industry. DURAN™ PURE Bottles are closed with a protective cover to prevent contamination. Very uniform, durable and chemically resistant amber colouring due to the use of innovative technology offers protection for light sensitive compounds. DWK Life Sciences provides CoAs with each delivery, assures 100% traceability and change control management. DURAN™ PURE Bottles are available with the documentation required by regulatory authorities and meet the requirements of the international pharmacopoeias.
  • Protective cover prevents contamination inside the bottle during storage and transport
  • Single batch delivery
  • Certificate of Analysis (CoA)
  • 100% visual inspection
  • Analysis of hydrolytic resistance according to EP, USP and JP for each batch
  • Change control management and information of customer in case of changes
  • 100% traceability of all manufacturing steps
  • Drug Master File
Specifications
Specifications
Manufacturing of light-sensitive pharmaceutical drugs and biotech products; Storage and transport of light-sensitive APIs, bulk drug intermediates, ingredients and media
Borosilicate Glass (3.3)
Autoclavable
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DWK Life Sciences DURAN™ PURE Bottle, Amber, with DIN Thread