Learn More
DWK Life Sciences DURAN™ Reagent Bottle, Wide Neck, Amber, USP /EP (3.2.1) Light Transmission
Description
The classical stoppered reagent bottles are moulded from amber DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a wide conical ground joint neck and comply with ISO 4796-2. The wider neck is more convenient for handling powders. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel. The amber glass ensures that light sensitive contents are protected from the effects of photo-degradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle.
- USP/EP UV protection up to approx. 500 nm due to external colouration
- The amber colour is only applied externally ensuring that internal surfaces have standard DURAN™ clear Type 1 glass properties
- Very durable and chemically resistant amber colour
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- All glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
Specifications
Diameter (Metric) | 133 mm |
Capacity (Metric) | 2000 mL |
Description | Neck with standard ground joint and glass flat-headed stopper |
For Use With (Application) | Storage of light sensitive reagents, pharmaceuticals, and chemicals; Transport and shipping of light sensitive samples; Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |