Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Reduction from sleeve to core.
Brand:  DWK Life Sciences 286548201
 View more versions of this product
Product Code. 10223881
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
  • Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Socket NS 29/32, Cone NS 24/29