Learn More
DWK Life Sciences DURAN™ Reduction Adapter, socket/cone
Reduction from sleeve to core.
Brand:  DWK Life Sciences 286535906
 View more versions of this product
Product Code. 15961601
  • £126.00 / Pack of 10
Estimated Shipment: 01-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Adapters are used to connect two components with different NS sizes. DURAN™ reducers allow the connection of a larger sleeve to a smaller core.
  • Reducers allow the connection of a larger sleeve to a smaller core and thus the use of differently sized ground joint components.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
Connection of two components with different standard ground joint sizes
Socket NS 19/26, Cone NS 14/23