missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Replacement Parts for HPLC screw cap GL 45
GREENER_CHOICE

Product Code. 10595263 Shop All DWK Life Sciences Products
Click to view available options
:
3.2 mm and blind silicone seals
This item is not returnable. View return policy

Product Code. 10595263

Brand: DWK Life Sciences 1129813

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pressure and vacuum resistant bottle for HPLC applications.

The DURAN™ HPLC bottle is available as a ready-made system. This comprises a special pressure and vacuum resistant bottle and a screw cap with 4 ports. This enables 3 stations to be served at the same time from one vessel. At the same time a sterile pressure equalisation can be fitted to the fourth outlet.

  • Connection of different hose diameters (1.6 and 3.2 mm)
  • Unused ports can be sealed with silicone blanking seals
  • Optional sterile pressure equalisation sets (membrane filter 0.2 μm
  • Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals available
  • Unused ports can be sealed off with a blank GL 18 screw cap
  • Fully compatible with all the GLS 80 threads and therefore highly versatile
TRUSTED_SUSTAINABILITY

Specifications

Description Replacement parts for HPLC screw cap (includes M8 caps and 1.6, 3.2 mm and blind silicone seals)
For Use With (Application) Pressure and vacuum resistant bottle for HPLC applications; Safe transfer of liquid media within a closed and sterile system; Supply of solvents to HPLC equipment
Material Borosilicate Glass (3.3)
Quantity 1 Ea.
Thread Size (Metric) GL 45
Closure Material Polypropylene with Silicone Seal
Autoclavable Autoclavable
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.