Learn More
DWK Life Sciences DURAN™ Round Bottom Flask, wide neck, with beaded rim
Due to their spherical shape, round bottom flasks are used for heating liquids, among other things.
£77.20 - £401.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | Suitable for heating mantles; Heating; Mixing several components; Sizes 50, 1000, 2000, 3000, 5000, 6000, 10000, 12000 and 20000 mL non-DIN ISO size |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11736093
|
DWK Life Sciences
217412407 |
100 mL |
£77.20
Pack of 10 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
11746093
|
DWK Life Sciences
217413609 |
250 mL |
£183.00
Pack of 10 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||||
11756093
|
DWK Life Sciences
217414408 |
500 mL |
£198.00
Pack of 10 |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
11776093
|
DWK Life Sciences
217416306 |
2000 mL |
£401.00
Pack of 10 |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
10517731
|
DWK Life Sciences
217417105 |
4000 mL |
£89.15
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
10130301
|
DWK Life Sciences
217417602 |
6000 mL |
£118.00
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
11851000
|
DWK Life Sciences
217415404 |
1000 mL |
£181.00
Pack of 10 |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
Description
Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.
- The wide neck enables easy filling and cleaning
- The geometry permits very uniform heating
- Fired-on, highly durable white ceramic
Specifications
Borosilicate Glass (3.3) | |
Suitable for heating mantles; Heating; Mixing several components; Sizes 50, 1000, 2000, 3000, 5000, 6000, 10000, 12000 and 20000 mL non-DIN ISO size | |
Autoclavable |